missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Methoxychalcone, 98%, ACROS Organics™

Chemical Identifiers

CAS 959-33-1
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Molecular Formula C16H14O2
Formula Weight 238.29
MDL Number MFCD00017179
Chemical Name or Material 4-Methoxychalcone
Assay ≥97.5%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
AC327640050 SDS Thermo Fisher Scientific
AC327640050
Glass bottle 5g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC327640250 SDS Thermo Fisher Scientific
327640250
Glass bottle 25g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

959-33-1
100.0
C16H14O2
MFCD00017179
≥97.5%
97.5
98%
238.29
4-Methoxychalcone

Specifications

Yellow
5g
97.5
Crystalline Powder
C16H14O2
MFCD00017179
4-Methoxychalcone
73° to 78°C
959-33-1
100.0
98%
238.29
Glass bottle
≥97.5%

Safety and Handling

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

EINECSNumber : 213-499-7