Learn More
4'-Methoxychalcone, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals B2215806
Description
4-Methoxychalcone is used as a precursor to prepare 3-(4-methoxy-phenyl)-3-morpholin-4-yl-1-phenyl-propan-1-one by reacting with morpholine in presence of heptanes as solvent.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications4-Methoxychalcone is used as a precursor to prepare 3-(4-methoxy-phenyl)-3-morpholin-4-yl-1-phenyl-propan-1-one by reacting with morpholine in presence of heptanes as solvent.
Solubility
Soluble in dichloromethane and methanol. Insoluble in water.
Notes
Keep the container tightly closed in a dry and well-ventilated place. Incompatible withstrong oxidizing agents.
Specifications
4'-Methoxychalcone | |
101°C to 103°C | |
MFCD00008407 | |
644640 | |
Soluble in dichloromethane and methanol. Insoluble in water. | |
COC1=CC=C(C=C1)C(=O)C=CC2=CC=CC=C2 | |
238.286 | |
238.29 |
959-23-9 | |
C16H14O2 | |
5 g | |
4'-methoxychalcone, 2e-1-4-methoxyphenyl-3-phenylprop-2-en-1-one, 1-4-methoxyphenyl-3-phenylprop-2-en-1-one, chembl34398, 1-4-methoxyphenyl-3-phenyl-2-propen-1-one, e-1-4-methoxyphenyl-3-phenylprop-2-en-1-one, unii-bm45n45fiz, e-1-4-methoxy-phenyl-3-phenyl-propenone, 2-propen-1-one, 1-4-methoxyphenyl-3-phenyl-, 2e, 2-propen-1-one, 1-4-methoxyphenyl-3-phenyl | |
KJHHAPASNNVTSN-KPKJPENVSA-N | |
(E)-1-(4-methoxyphenyl)-3-phenylprop-2-en-1-one | |
641818 | |
97% |
Chemical Identifiers
959-23-9 | |
238.286 | |
KJHHAPASNNVTSN-KPKJPENVSA-N | |
641818 | |
COC1=CC=C(C=C1)C(=O)C=CC2=CC=CC=C2 |
C16H14O2 | |
MFCD00008407 | |
4'-methoxychalcone, 2e-1-4-methoxyphenyl-3-phenylprop-2-en-1-one, 1-4-methoxyphenyl-3-phenylprop-2-en-1-one, chembl34398, 1-4-methoxyphenyl-3-phenyl-2-propen-1-one, e-1-4-methoxyphenyl-3-phenylprop-2-en-1-one, unii-bm45n45fiz, e-1-4-methoxy-phenyl-3-phenyl-propenone, 2-propen-1-one, 1-4-methoxyphenyl-3-phenyl-, 2e, 2-propen-1-one, 1-4-methoxyphenyl-3-phenyl | |
(E)-1-(4-methoxyphenyl)-3-phenylprop-2-en-1-one |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 213-495-5
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only