missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-(Methoxycarbonyl)bicyclo[2.2.2]octan-1-carboxylic Acid, TCI America™

$131.39 - $131.39

Chemical Identifiers

CAS 18720-35-9
Molecular Formula C11H16O4
Molecular Weight (g/mol) 212.245
MDL Number MFCD18459359
InChI Key KBZVAQVEHVGIEI-UHFFFAOYSA-N
Synonym Monomethyl Hydrogen Bicyclo[2.2.2]octane-1,4-dicarboxylate, Bicyclo[2.2.2]octane-1,4-dicarboxylic Acid Monomethyl Ester
PubChem CID 11042062
IUPAC Name 4-methoxycarbonylbicyclo[2.2.2]octane-1-carboxylic acid
SMILES COC(=O)C12CCC(CC1)(CC2)C(=O)O
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M28551G View Documents TCI America
M28551G
1g Each for $131.39
 
Specifications

Chemical Identifiers

18720-35-9
212.245
KBZVAQVEHVGIEI-UHFFFAOYSA-N
11042062
COC(=O)C12CCC(CC1)(CC2)C(=O)O
C11H16O4
MFCD18459359
Monomethyl Hydrogen Bicyclo[2.2.2]octane-1,4-dicarboxylate, Bicyclo[2.2.2]octane-1,4-dicarboxylic Acid Monomethyl Ester
4-methoxycarbonylbicyclo[2.2.2]octane-1-carboxylic acid

Specifications

White
1g
C11H16O4
Monomethyl Hydrogen Bicyclo[2.2.2]octane-1,4-dicarboxylate, Bicyclo[2.2.2]octane-1,4-dicarboxylic Acid Monomethyl Ester
COC(=O)C12CCC(CC1)(CC2)C(=O)O
212.245
212.25
≥98.0% (GC,T)
188°C
18720-35-9
MFCD18459359
KBZVAQVEHVGIEI-UHFFFAOYSA-N
4-methoxycarbonylbicyclo[2.2.2]octane-1-carboxylic acid
11042062
Crystal-Powder at 20°C
4-(Methoxycarbonyl)bicyclo[2.2.2]octan-1-carboxylic Acid
Safety and Handling

Safety and Handling

TSCA : No