missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Methoxycarbonyl-3-chlorobenzeneboronic acid pinacol ester, 98%

Manufacturer:  Alfa Aesar™ H53331MD

 View more versions of this product

Catalog No. AAH53331MD



Description

Description

Applications
4-Methoxycarbonyl-3-chlorobenzeneboronic acid pinacol ester is used as organic reagents.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Specifications

Specifications

4-Methoxycarbonyl-3-chlorobenzeneboronic acid pinacol ester
250mg
C14H18BClO4
methyl 2-chloro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzoate, 3-chloro-4-methoxycarbonylphenylboronic acid, pinacol ester, 3-chloro-4-methoxycarbonylphenylboronic acid pinacol ester, 3-chloro-4-methoxycarbonyl benzeneboronic acid pinacol ester, 3-chloro-4-methoxycarbonyl benzeneboronic acid, pinacol ester, methyl 2-chloro-4-tetramethyl-1,3,2-dioxaborolan-2-yl benzoate, 4-methoxycarbonyl-3-chlorophenylboronic acid, pinacol ester, methyl 2-chloro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolane-2-yl benzoate, methyl2-chloro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzoate, 2-3-chloro-4-methoxycarbonyl phenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
B1(OC(C(O1)(C)C)(C)C)C2=CC(=C(C=C2)C(=O)OC)Cl
296.554
296.56
48°C to 51°C
334018-52-9
MFCD06659928
SPVAFOQLYCMOAO-UHFFFAOYSA-N
methyl 2-chloro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoate
22493932
98%

Chemical Identifiers

334018-52-9
296.554
SPVAFOQLYCMOAO-UHFFFAOYSA-N
22493932
B1(OC(C(O1)(C)C)(C)C)C2=CC(=C(C=C2)C(=O)OC)Cl
C14H18BClO4
MFCD06659928
methyl 2-chloro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzoate, 3-chloro-4-methoxycarbonylphenylboronic acid, pinacol ester, 3-chloro-4-methoxycarbonylphenylboronic acid pinacol ester, 3-chloro-4-methoxycarbonyl benzeneboronic acid pinacol ester, 3-chloro-4-methoxycarbonyl benzeneboronic acid, pinacol ester, methyl 2-chloro-4-tetramethyl-1,3,2-dioxaborolan-2-yl benzoate, 4-methoxycarbonyl-3-chlorophenylboronic acid, pinacol ester, methyl 2-chloro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolane-2-yl benzoate, methyl2-chloro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzoate, 2-3-chloro-4-methoxycarbonyl phenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
methyl 2-chloro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoate
SDS Documents