4'-Methoxybiphenyl-4-sulfonyl chloride, 95%, Acros Organics™
Manufacturer:  Acros Organics AC450400010
 View more versions of this product
Catalog No. AC450400010
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications
Specifications
4'-Methoxybiphenyl-4-sulfonyl chloride
104.0°C to 109.0°C
202752-04-3
4'-methoxybiphenyl-4-sulfonyl chloride, 4'-methoxy 1,1'-biphenyl-4-sulfonyl chloride, 4-4-methoxyphenyl benzenesulfonyl chloride, 4'-methoxy-biphenyl-4-sulfonyl chloride, 4'-methoxy-1,1'-biphenyl-4-sulfonyl chloride, 4-4-methoxyphenyl phenyl sulfonyl chloride, 1,1'-biphenyl-4-sulfonylchloride, 4'-methoxy, 4'-methoxy biphenyl-4-sulfonyl chloride, 4-4-methoxyphenyl benzenesulfonylchloride, 4-4-methoxyphenyl phenyl sulfonylchloride
COC1=CC=C(C=C1)C2=CC=C(C=C2)S(=O)(=O)Cl
282.738
282.75
Authentic
1g
C13H11ClO3S
AJWXJCPSCXFPJJ-UHFFFAOYSA-N
4-(4-methoxyphenyl)benzenesulfonyl chloride
2794750
95%
Chemical Identifiers
202752-04-3
282.738
4'-methoxybiphenyl-4-sulfonyl chloride, 4'-methoxy 1,1'-biphenyl-4-sulfonyl chloride, 4-4-methoxyphenyl benzenesulfonyl chloride, 4'-methoxy-biphenyl-4-sulfonyl chloride, 4'-methoxy-1,1'-biphenyl-4-sulfonyl chloride, 4-4-methoxyphenyl phenyl sulfonyl chloride, 1,1'-biphenyl-4-sulfonylchloride, 4'-methoxy, 4'-methoxy biphenyl-4-sulfonyl chloride, 4-4-methoxyphenyl benzenesulfonylchloride, 4-4-methoxyphenyl phenyl sulfonylchloride
4-(4-methoxyphenyl)benzenesulfonyl chloride
C13H11ClO3S
AJWXJCPSCXFPJJ-UHFFFAOYSA-N
2794750
COC1=CC=C(C=C1)C2=CC=C(C=C2)S(=O)(=O)Cl
Safety and Handling
Safety and Handling
GHS H Statement Causes severe skin burns and eye damage. Contact with water liberates toxic gas.
GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. IF SWALLOWED: rinse mouth. Do NOT induce vomiting. Immediately call a POISON CENTER or doctor/physician. IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Store in a dry place. Store in a closed container.
Danger
SDS Documents