Please login to your online account to display your discounted pricing

Alfa Aesar™ 4'-Methoxybiphenyl-4-boronic acid pinacol ester, 95%

Chemical Identifiers

Chemical Name or Material 4'-Methoxybiphenyl-4-boronic acid pinacol ester
Molecular Formula C19H23BO3
Formula Weight 310.2
MDL Number MFCD16294530
Synonym 2-(4'-Methoxy-4-biphenylyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane; 4-(4-Methoxyphenyl)benzeneboronic acid pinacol ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5190703

 
alfa aesar™
H5190703
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH5190706

 
alfa aesar™
H5190706
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 310.2
Chemical Name or Material 4'-Methoxybiphenyl-4-boronic acid pinacol ester
Assay 95%
Molecular Formula C19H23BO3
MDL Number MFCD16294530
Synonym 2-(4'-Methoxy-4-biphenylyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane; 4-(4-Methoxyphenyl)benzeneboronic acid pinacol ester
Quantity 1g