Please login to your online account to display your discounted pricing

Alfa Aesar™ (4-Methoxybenzylidene)malononitrile, 99%


Quantity 100mg

Manufacturer: alfa aesar™  J67456MC

 View more versions of this product

Catalog No. AAJ67456MC

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 184.2
Melting Point 166° to 168°C
Chemical Name or Material (4-Methoxybenzylidene)malononitrile
Physical Form Solid
MDL Number MFCD00019787
CAS 2826-26-8
Health Hazard 2 GHS H Statement
H302-H312-H332-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Color Yellow
Assay 99%
UN Number UN3439
Sensitivity Light Sensitive
Health Hazard 1 Warning
DOT Information Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
Synonym Tyrphostin 1
Quantity 100mg

Specifications

Chemical Name or Material (4-Methoxybenzylidene)malononitrile
CAS 2826-26-8
Formula Weight 184.2
MDL Number MFCD00019787
UN Number UN3439
Synonym Tyrphostin 1
Accessories

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H302-H312-H332-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.