Learn More
4-Methoxybenzylchloride, 98%, stabilized, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 294170250
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
4-Methoxybenzylchloride | |
824-94-2,584-08-7 | |
100.0 | |
Colorless to Yellow | |
117.0°C to 118.0°C (14.0 mmHg) | |
Authentic | |
Glass bottle | |
1.5470 to 1.5490 | |
25 g | |
4-methoxybenzyl chloride, 4-methoxybenzylchloride, 1-chloromethyl-4-methoxybenzene, p-methoxybenzyl chloride, p-anisyl chloride, alpha-chloro-4-methoxytoluene, p-chloromethyl anisole, benzene, 1-chloromethyl-4-methoxy, 4-chloromethyl anisole, anisole, p-chloromethyl | |
MOHYOXXOKFQHDC-UHFFFAOYSA-N | |
1-(chloromethyl)-4-methoxybenzene | |
1% K2CO3 | |
156.61 | |
Liquid |
98% | |
97.5 | |
-1.0°C | |
1.1500g/mL | |
109°C | |
97.5% min. (GC) | |
C8H9ClO | |
MFCD00000915 | |
1.15 | |
Solubility in water: slowly decomposes | |
COC1=CC=C(C=C1)CCl | |
156.61 | |
69993 | |
98% |
Chemical Identifiers
824-94-2 | |
156.61 | |
MOHYOXXOKFQHDC-UHFFFAOYSA-N | |
69993 | |
COC1=CC=C(C=C1)CCl |
C8H9ClO | |
MFCD00000915 | |
4-methoxybenzyl chloride, 4-methoxybenzylchloride, 1-chloromethyl-4-methoxybenzene, p-methoxybenzyl chloride, p-anisyl chloride, alpha-chloro-4-methoxytoluene, p-chloromethyl anisole, benzene, 1-chloromethyl-4-methoxy, 4-chloromethyl anisole, anisole, p-chloromethyl | |
1-(chloromethyl)-4-methoxybenzene |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
GHS Signal Word: Danger
EINECSNumber : 212-540-6
RUO – Research Use Only