missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Methoxybenzylamine, TCI America™

Manufacturer:  TCI America M0870250ML

 View more versions of this product

Catalog No. M0870250ML



Specifications

Specifications

4-Methoxybenzylamine
Yellow
2393-23-9
MFCD00008122
4-methoxybenzylamine, 4-methoxyphenyl methanamine, benzenemethanamine, 4-methoxy, p-methoxybenzylamine, 4-methoxy-benzylamine, anisylamine, benzylamine, p-methoxy, 4-methoxybenzyl amine, 1-4-methoxyphenyl methanamine, 4-methoxyphenyl methylamine
COC1=CC=C(C=C1)CN
137.182
CHEBI:49837
Clear Liquid at 20°C
236°C
250mL
C8H11NO
2735
IDPURXSQCKYKIJ-UHFFFAOYSA-N
(4-methoxyphenyl)methanamine
75452
137.18
≥97.0% (GC,T)

Chemical Identifiers

2393-23-9
137.182
IDPURXSQCKYKIJ-UHFFFAOYSA-N
75452
(4-methoxyphenyl)methanamine
C8H11NO
MFCD00008122
4-methoxybenzylamine, 4-methoxyphenyl methanamine, benzenemethanamine, 4-methoxy, p-methoxybenzylamine, 4-methoxy-benzylamine, anisylamine, benzylamine, p-methoxy, 4-methoxybenzyl amine, 1-4-methoxyphenyl methanamine, 4-methoxyphenyl methylamine
CHEBI:49837
COC1=CC=C(C=C1)CN
Safety and Handling

Safety and Handling

TSCA : Yes

Documents