missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
4-Methoxybenzylamine, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 125780250
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
4-Methoxybenzylamine | |
Colorless to Yellow | |
236.0°C to 237.0°C | |
Authentic | |
Glass bottle | |
1.5460 to 1.5480 | |
25 mL | |
1.05 | |
IDPURXSQCKYKIJ-UHFFFAOYSA-N | |
(4-methoxyphenyl)methanamine | |
75452 | |
137.18 | |
Liquid |
2393-23-9 | |
1.0500g/mL | |
>110°C | |
97.5% min. (GC) | |
C8H11NO | |
CH3OC6H4CH2NH2 | |
13,606 | |
4-methoxybenzylamine, 4-methoxyphenyl methanamine, benzenemethanamine, 4-methoxy, p-methoxybenzylamine, 4-methoxy-benzylamine, anisylamine, benzylamine, p-methoxy, 4-methoxybenzyl amine, 1-4-methoxyphenyl methanamine, 4-methoxyphenyl methylamine | |
COC1=CC=C(C=C1)CN | |
137.18 | |
CHEBI:49837 | |
98% |
Chemical Identifiers
2393-23-9 | |
137.18 | |
4-methoxybenzylamine, 4-methoxyphenyl methanamine, benzenemethanamine, 4-methoxy, p-methoxybenzylamine, 4-methoxy-benzylamine, anisylamine, benzylamine, p-methoxy, 4-methoxybenzyl amine, 1-4-methoxyphenyl methanamine, 4-methoxyphenyl methylamine | |
CHEBI:49837 | |
COC1=CC=C(C=C1)CN |
C8H11NO | |
IDPURXSQCKYKIJ-UHFFFAOYSA-N | |
75452 | |
(4-methoxyphenyl)methanamine |
Safety and Handling
GHS Signal Word: Danger
EINECSNumber : 219-247-2
RUO – Research Use Only