Learn More
4-Methoxybenzylamine, 98+%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A1499914
Description
4-Methoxybenzylamine is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuff. It is also used fine chemical intemediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications4-Methoxybenzylamine is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuff. It is also used fine chemical intemediate.
Solubility
It is highly soluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Moisture sensitive. Store under inert gas. Keep away from Strong oxidizing agentsacids, Acid chlorides, Acid anhydrides, Oxidizing agents, Chloroformates.
Specifications
4-Methoxybenzylamine | |
-10°C | |
236°C to 237°C | |
Amine-like | |
1.546 | |
25 g | |
508206 | |
4-methoxybenzylamine, 4-methoxyphenyl methanamine, benzenemethanamine, 4-methoxy, p-methoxybenzylamine, 4-methoxy-benzylamine, anisylamine, benzylamine, p-methoxy, 4-methoxybenzyl amine, 1-4-methoxyphenyl methanamine, 4-methoxyphenyl methylamine | |
IDPURXSQCKYKIJ-UHFFFAOYSA-N | |
(4-methoxyphenyl)methanamine | |
75452 | |
137.18 |
2393-23-9 | |
1.053 | |
114°C (237°F) | |
C8H11NO | |
MFCD00008122 | |
UN2735 | |
Air sensitive; May form a precipitate | |
It is highly soluble in water. | |
COC1=CC=C(C=C1)CN | |
137.182 | |
CHEBI:49837 | |
≥98% |
Chemical Identifiers
2393-23-9 | |
137.182 | |
IDPURXSQCKYKIJ-UHFFFAOYSA-N | |
75452 | |
(4-methoxyphenyl)methanamine |
C8H11NO | |
MFCD00008122 | |
4-methoxybenzylamine, 4-methoxyphenyl methanamine, benzenemethanamine, 4-methoxy, p-methoxybenzylamine, 4-methoxy-benzylamine, anisylamine, benzylamine, p-methoxy, 4-methoxybenzyl amine, 1-4-methoxyphenyl methanamine, 4-methoxyphenyl methylamine | |
CHEBI:49837 | |
COC1=CC=C(C=C1)CN |
Safety and Handling
GHS H Statement
H314-H318-H302
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314
DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.
EINECSNumber : 219-247-2
TSCA : Yes
Recommended Storage : Ambient temperatures; Store under Argon
RUO – Research Use Only