missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Methoxybenzyl isocyanate, 99%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC319400050

 View more versions of this product

Catalog No. AC319400050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

4-Methoxybenzyl isocyanate
1.5305 to 1.5325
5g
98.5
99%
MFCD00673064
QRBHVARIMDDOOV-UHFFFAOYSA-N
1-(isocyanatomethyl)-4-methoxybenzene
187910
99%
Glass bottle
90.0°C (0.6 mmHg)
56651-60-6
100.0
C9H9NO2
1-isocyanatomethyl-4-methoxybenzene, 4-methoxybenzyl isocyanate, 4-methoxybenzylisocyanate, 4-methoxyphenyl methanisocyanate, acmc-20amf5, benzene, 1-isocyanatomethyl-4-methoxy, isocyanic acid, p-methoxybenzyl, p-methoxybenzyl isocyanate, p-methoxybenzylisocyanate, p-metoxybenzyl-isocyanate
COC1=CC=C(C=C1)CN=C=O
163.18
163.18

Chemical Identifiers

56651-60-6
163.18
QRBHVARIMDDOOV-UHFFFAOYSA-N
187910
COC1=CC=C(C=C1)CN=C=O
C9H9NO2
MFCD00673064
1-isocyanatomethyl-4-methoxybenzene, 4-methoxybenzyl isocyanate, 4-methoxybenzylisocyanate, 4-methoxyphenyl methanisocyanate, acmc-20amf5, benzene, 1-isocyanatomethyl-4-methoxy, isocyanic acid, p-methoxybenzyl, p-methoxybenzyl isocyanate, p-methoxybenzylisocyanate, p-metoxybenzyl-isocyanate
1-(isocyanatomethyl)-4-methoxybenzene
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if inhaled.
Harmful in contact with skin.
Harmful if swallowed.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

SDS Documents