missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Methoxybenzyl Alcohol, TCI America™

Manufacturer:  TCI America M0107500G

 View more versions of this product

Catalog No. M0107500G



Specifications

Specifications

4-Methoxybenzyl Alcohol
White
105-13-5
MFCD00004653
MSHFRERJPWKJFX-UHFFFAOYSA-N
(4-methoxyphenyl)methanol
7738
138.17
≥98.0% (GC)
259°C
500g
C8H10O2
4-methoxybenzyl alcohol, 4-methoxyphenyl methanol, anise alcohol, anisyl alcohol, p-anisyl alcohol, p-methoxybenzyl alcohol, benzenemethanol, 4-methoxy, anisic alcohol, p-anisol alcohol, 4-methoxybenzenemethanol
COC1=CC=C(C=C1)CO
138.166
CHEBI:86918
Crystal-Lump at 20°C

Chemical Identifiers

105-13-5
138.166
MSHFRERJPWKJFX-UHFFFAOYSA-N
7738
(4-methoxyphenyl)methanol
C8H10O2
MFCD00004653
4-methoxybenzyl alcohol, 4-methoxyphenyl methanol, anise alcohol, anisyl alcohol, p-anisyl alcohol, p-methoxybenzyl alcohol, benzenemethanol, 4-methoxy, anisic alcohol, p-anisol alcohol, 4-methoxybenzenemethanol
CHEBI:86918
COC1=CC=C(C=C1)CO
Safety and Handling

Safety and Handling

EINECSNumber : (3)-1054

RTECSNumber : DO8925000

TSCA : Yes

Documents