missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Methoxybenzyl 2,2,2-Trichloroacetimidate, TCI America™

$155.84 - $580.56

Chemical Identifiers

CAS 89238-99-3
Molecular Formula C10H10Cl3NO2
Molecular Weight (g/mol) 282.545
MDL Number MFCD00134547
InChI Key TYHGKLBJBHACOI-UHFFFAOYSA-N
Synonym 2,2,2-Trichloroacetimidic Acid 4-Methoxybenzyl Ester
PubChem CID 11087263
IUPAC Name (4-methoxyphenyl)methyl 2,2,2-trichloroethanimidate
SMILES COC1=CC=C(C=C1)COC(=N)C(Cl)(Cl)Cl
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M20165G View Documents TCI America
M20165G
5g Each for $155.84
 
M201625G View Documents TCI America
M201625G
25g Each for $580.56
 
Specifications

Chemical Identifiers

89238-99-3
282.545
TYHGKLBJBHACOI-UHFFFAOYSA-N
11087263
COC1=CC=C(C=C1)COC(=N)C(Cl)(Cl)Cl
C10H10Cl3NO2
MFCD00134547
2,2,2-Trichloroacetimidic Acid 4-Methoxybenzyl Ester
(4-methoxyphenyl)methyl 2,2,2-trichloroethanimidate

Specifications

137°C
5g
C10H10Cl3NO2
2,2,2-Trichloroacetimidic Acid 4-Methoxybenzyl Ester
COC1=CC=C(C=C1)COC(=N)C(Cl)(Cl)Cl
282.545
282.55
≥96.0% (GC)
Yellow
89238-99-3
MFCD00134547
TYHGKLBJBHACOI-UHFFFAOYSA-N
(4-methoxyphenyl)methyl 2,2,2-trichloroethanimidate
11087263
Clear Liquid at 20°C
4-Methoxybenzyl 2,2,2-Trichloroacetimidate
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Freezer