missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Methoxybenzoyl Chloride, TCI America™

Manufacturer:  TCI America M0721100G

 View more versions of this product

Catalog No. M0721100G



Specifications

Specifications

4-Methoxybenzoyl Chloride
100g
C8H7ClO2
1729
MXMOTZIXVICDSD-UHFFFAOYSA-N
4-methoxybenzoyl chloride
7477
Crystal-Lump at 20°C
White-Yellow
100-07-2
MFCD00000687
p-anisoyl chloride, benzoyl chloride, 4-methoxy, anisoyl chloride, 4-anisoyl chloride, p-methoxybenzoyl chloride, 4-methoxybenzoylchloride, 4-methoxybenzoic acid chloride, benzoyl chloride, p-methoxy, p-methoxybenzoic acid chloride, 4-methoxy-benzoyl chloride
COC1=CC=C(C=C1)C(=O)Cl
170.592
170.59
≥99.0% (GC)

Chemical Identifiers

100-07-2
170.592
MXMOTZIXVICDSD-UHFFFAOYSA-N
7477
COC1=CC=C(C=C1)C(=O)Cl
C8H7ClO2
MFCD00000687
p-anisoyl chloride, benzoyl chloride, 4-methoxy, anisoyl chloride, 4-anisoyl chloride, p-methoxybenzoyl chloride, 4-methoxybenzoylchloride, 4-methoxybenzoic acid chloride, benzoyl chloride, p-methoxy, p-methoxybenzoic acid chloride, 4-methoxy-benzoyl chloride
4-methoxybenzoyl chloride
Safety and Handling

Safety and Handling

EINECSNumber : (3)-1684

RTECSNumber : CA0270000

TSCA : Yes

Recommended Storage : Refrigerator

Documents