Learn More
4-Methoxybenzoyl chloride, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals L1312022
Description
4-Methoxybenzoyl chloride is used in the synthesis of stilbene and dihydrostilbene derivatives as potential anti-cancer agents. It is also used in the synthesis of coumarin dimers with potential HIV-1 activity.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications4-Methoxybenzoyl chloride is used in the synthesis of stilbene and dihydrostilbene derivatives as potential anti-cancer agents. It is also used in the synthesis of coumarin dimers with potential HIV-1 activity.
Solubility
It reacts in water.
Notes
Moisture sensitive. Incompatible with oxidizing agents and bases.
Specifications
4-Methoxybenzoyl chloride | |
22°C | |
145°C (14 mmHg) | |
C8H7ClO2 | |
MFCD00000687 | |
UN1729 | |
Moisture sensitive | |
p-anisoyl chloride, benzoyl chloride, 4-methoxy, anisoyl chloride, 4-anisoyl chloride, p-methoxybenzoyl chloride, 4-methoxybenzoylchloride, 4-methoxybenzoic acid chloride, benzoyl chloride, p-methoxy, p-methoxybenzoic acid chloride, 4-methoxy-benzoyl chloride | |
MXMOTZIXVICDSD-UHFFFAOYSA-N | |
4-methoxybenzoyl chloride | |
7477 | |
97% |
100-07-2 | |
1.26 | |
87°C (188°F) | |
1.581 | |
100 g | |
471918 | |
14,673 | |
It reacts in water. | |
COC1=CC=C(C=C1)C(Cl)=O | |
170.59 | |
170.6 |
Chemical Identifiers
100-07-2 | |
170.59 | |
MXMOTZIXVICDSD-UHFFFAOYSA-N | |
7477 | |
COC1=CC=C(C=C1)C(Cl)=O |
C8H7ClO2 | |
MFCD00000687 | |
p-anisoyl chloride, benzoyl chloride, 4-methoxy, anisoyl chloride, 4-anisoyl chloride, p-methoxybenzoyl chloride, 4-methoxybenzoylchloride, 4-methoxybenzoic acid chloride, benzoyl chloride, p-methoxy, p-methoxybenzoic acid chloride, 4-methoxy-benzoyl chloride | |
4-methoxybenzoyl chloride |
Safety and Handling
GHS H Statement
H314-H318-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.
P210-P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
H227-H314
DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: ANISOYL CHLORIDE
EINECSNumber : 202-816-4
TSCA : Yes
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only