missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Methoxybenzophenone, 98+%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals B2312314

 View more versions of this product

Catalog No. AAB2312314



Description

Description

4-Methoxybenzophenone and its analogues showed diverse plant growth-regulating actions such as inhibition of shoot and root growth, induction of chlorosis, and a disturbance in phototropism or geotropism. It is also used as a photo polymerization catalyst.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Methoxybenzophenone and its analogues showed diverse plant growth-regulating actions such as inhibition of shoot and root growth, induction of chlorosis, and a disturbance in phototropism or geotropism. It is also used as a photo polymerization catalyst.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agent.
Specifications

Specifications

4-Methoxybenzophenone
60°C to 63°C
C14H12O2
25 g
4-methoxybenzophenone, 4-methoxyphenyl phenyl methanone, p-methoxybenzophenone, benzophenone, 4-methoxy, methanone, 4-methoxyphenyl phenyl, phenyl p-anisyl ketone, 4-benzoylanisole, 4-methoxyphenyl phenylmethanone, unii-i4xj07373m, 4-methoxyphenyl-phenylmethanone
SWFHGTMLYIBPPA-UHFFFAOYSA-N
(4-methoxyphenyl)-phenylmethanone
69146
≥98%
611-94-9
354°C to 356°C
MFCD00008403
1104713
Insoluble in water.
COC1=CC=C(C=C1)C(=O)C2=CC=CC=C2
212.248
212.25

Chemical Identifiers

611-94-9
212.248
SWFHGTMLYIBPPA-UHFFFAOYSA-N
69146
COC1=CC=C(C=C1)C(=O)C2=CC=CC=C2
C14H12O2
MFCD00008403
4-methoxybenzophenone, 4-methoxyphenyl phenyl methanone, p-methoxybenzophenone, benzophenone, 4-methoxy, methanone, 4-methoxyphenyl phenyl, phenyl p-anisyl ketone, 4-benzoylanisole, 4-methoxyphenyl phenylmethanone, unii-i4xj07373m, 4-methoxyphenyl-phenylmethanone
(4-methoxyphenyl)-phenylmethanone
Safety and Handling

Safety and Handling

EINECSNumber : 210-285-5

RTECSNumber : PC4962500

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only