missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
4-Methoxybenzophenone, 97%, ACROS Organics™
Manufacturer: Thermo Fisher Scientific 125741000
Specifications
4-Methoxybenzophenone | |
Glass bottle | |
Orange-Yellow to White | |
100g | |
611-94-9 | |
100.0 | |
C14H12O2 | |
MFCD00008403 | |
4-benzoylanisole, 4-methoxybenzophenone, 4-methoxyphenyl phenyl methanone, 4-methoxyphenyl phenylmethanone, 4-methoxyphenyl-phenylmethanone, benzophenone, 4-methoxy, methanone, 4-methoxyphenyl phenyl, p-methoxybenzophenone, phenyl p-anisyl ketone, unii-i4xj07373m | |
SWFHGTMLYIBPPA-UHFFFAOYSA-N | |
(4-methoxyphenyl)-phenylmethanone | |
69146 | |
Crystalline Solid |
Authentic | |
354°C to 356°C | |
58°C to 63°C | |
97% | |
96.0 | |
96% min. (GC) | |
CH3OC6H4COC6H5 | |
08, 159 | |
Solubility in water: insoluble in water. Other solubilities: soluble in ethanol and ether | |
COC1=CC=C(C=C1)C(=O)C2=CC=CC=C2 | |
212.25 | |
212.25 | |
97% |
Chemical Identifiers
611-94-9 | |
212.25 | |
SWFHGTMLYIBPPA-UHFFFAOYSA-N | |
69146 | |
COC1=CC=C(C=C1)C(=O)C2=CC=CC=C2 |
C14H12O2 | |
MFCD00008403 | |
4-benzoylanisole, 4-methoxybenzophenone, 4-methoxyphenyl phenyl methanone, 4-methoxyphenyl phenylmethanone, 4-methoxyphenyl-phenylmethanone, benzophenone, 4-methoxy, methanone, 4-methoxyphenyl phenyl, p-methoxybenzophenone, phenyl p-anisyl ketone, unii-i4xj07373m | |
(4-methoxyphenyl)-phenylmethanone |
Safety and Handling
EINECSNumber : 210-285-5
RTECSNumber : PC4962500