missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Methoxybenzophenone, 97%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 125740250

 View more versions of this product

Catalog No. AC125740250

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Specifications

4-Methoxybenzophenone
Glass bottle
Orange-Yellow to White
25g
611-94-9
100.0
C14H12O2
MFCD00008403
4-benzoylanisole, 4-methoxybenzophenone, 4-methoxyphenyl phenyl methanone, 4-methoxyphenyl phenylmethanone, 4-methoxyphenyl-phenylmethanone, benzophenone, 4-methoxy, methanone, 4-methoxyphenyl phenyl, p-methoxybenzophenone, phenyl p-anisyl ketone, unii-i4xj07373m
SWFHGTMLYIBPPA-UHFFFAOYSA-N
(4-methoxyphenyl)-phenylmethanone
69146
Crystalline Solid
Authentic
354°C to 356°C
58°C to 63°C
97%
96.0
96% min. (GC)
CH3OC6H4COC6H5
08, 159
Solubility in water: insoluble in water. Other solubilities: soluble in ethanol and ether
COC1=CC=C(C=C1)C(=O)C2=CC=CC=C2
212.25
212.25
97%

Chemical Identifiers

611-94-9
212.25
SWFHGTMLYIBPPA-UHFFFAOYSA-N
69146
COC1=CC=C(C=C1)C(=O)C2=CC=CC=C2
C14H12O2
MFCD00008403
4-benzoylanisole, 4-methoxybenzophenone, 4-methoxyphenyl phenyl methanone, 4-methoxyphenyl phenylmethanone, 4-methoxyphenyl-phenylmethanone, benzophenone, 4-methoxy, methanone, 4-methoxyphenyl phenyl, p-methoxybenzophenone, phenyl p-anisyl ketone, unii-i4xj07373m
(4-methoxyphenyl)-phenylmethanone
Safety and Handling

Safety and Handling

EINECSNumber : 210-285-5

RTECSNumber : PC4962500

SDS Documents