missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Methoxybenzoic anhydride, 98%

Manufacturer:  Alfa Aesar™ L0618014

 View more versions of this product

Catalog No. AAL0618014


 

Specifications

Description

4-Methoxybenzoic anhydride; 98%; Alfa Aesar; 25g; FW: 286.29; C16H14O5; MP: 95 to 99; CAS: 794-94-5

Applications
4-Methoxybenzoic anhydride is used as pharmaceutical intermediates.

Solubility
Soluble in alcohol, ether. Insoluble in water.

Notes
Moisture Sensitive. Protect from humidity and water. Incompatible with moisture and oxidizing agents.
Description & Specifications

Specifications

4-Methoxybenzoic anhydride
25g
98%
286.29
545712
95° to 99°C
794-94-5
C16H14O5
MFCD00017175
p-Anisic anhydride

Chemical Identifiers

794-94-5
286.29
545712
4-Methoxybenzoic anhydride
C16H14O5
MFCD00017175
p-Anisic anhydride

Safety and Handling

SDS & Safety Documents

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

RTECSNumber : BZ5300000