missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Methoxybenzensulfonic acid, 95%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC433120050

 View more versions of this product

Catalog No. AC433120050


 

Specifications Description & Specifications

Specifications

4-Methoxybenzensulfonic acid
5g
94.0
Authentic
≥94.0%
C7H8O4S
Glass bottle
White to pink
5857-42-1,7732-18-5
100.0
Crystalline Powder or Solid
95%
188.2
16% max.

Chemical Identifiers

5857-42-1,7732-18-5
100.0
C7H8O4S
4-Methoxybenzensulfonic acid
94.0
95%
188.2
≥94.0%

Safety and Handling

SDS & Safety Documents

GHS H Statement:
Causes severe skin burns and eye damage.
May cause an allergic skin reaction.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger