Please login to your online account to display your discounted pricing

4-Methoxybenzensulfonic acid, 95%, ACROS Organics™

$132.67 - $395.97

Chemical Identifiers

Chemical Name or Material 4-Methoxybenzensulfonic acid
CAS 5857-42-1
CAS Min % 94.0
CAS Max % 100.0
CAS 7732-18-5
Assay ≥94.0%
Assay Percent Range 95%
Molecular Formula C7H8O4S
Formula Weight 188.2
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC433120050

 
thermo fisher scientific
AC433120050
Glass bottle 5g Each for $132.67

AC433120250

 
thermo fisher scientific
AC433120250
Glass bottle 25g Each for $395.97
Description & Specifications

Specifications

Physical Form Crystalline Powder or Solid
Assay ≥94.0%
Formula Weight 188.2
Quantity 5g
Color White to pink
CAS Min % 94.0
CAS Max % 100.0
Chemical Name or Material 4-Methoxybenzensulfonic acid
Assay Percent Range 95%
Packaging Glass bottle
Water 16% max.
Health Hazard 2 GHS H Statement:
Causes severe skin burns and eye damage.
May cause an allergic skin reaction.
Infrared Spectrum Authentic
Molecular Formula C7H8O4S
CAS 5857-42-1
Health Hazard 1 GHS Signal Word: Danger
CAS 7732-18-5
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Safety and Handling

GHS Signal Word: Danger

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes severe skin burns and eye damage.
May cause an allergic skin reaction.