Learn More
4-Methoxybenzenesulfonyl Chloride, 99%, ACROS Organics™
Manufacturer: Thermo Fisher Scientific 125731000
Specifications
4-Methoxybenzenesulfonyl chloride | |
Authentic | |
Beige to White | |
100g | |
98.5 | |
98.5% min. (GC) | |
CH3OC6H4SO2Cl | |
11, 243 | |
4-methoxy-benzenesulfonyl chloride, 4-methoxybenzene-1-sulfonyl chloride, 4-methoxybenzenesulfonylchloride, 4-methoxybenzenesulphonyl chloride, 4-methoxyphenylsulfonyl chloride, benzenesulfonyl chloride, 4-methoxy, benzenesulfonyl chloride, p-methoxy, p-anisolesulfonyl chloride, p-anisylsulfonyl chloride, p-methoxyphenylsulfonyl chloride | |
DTJVECUKADWGMO-UHFFFAOYSA-N | |
4-methoxybenzenesulfonyl chloride | |
7401 | |
Crystalline Solid |
>110°C | |
Glass bottle | |
40.0°C to 42.0°C | |
98-68-0 | |
100.0 | |
C7H7ClO3S | |
MFCD00007446 | |
09,297 | |
Solubility in water: decomposes | |
COC1=CC=C(C=C1)S(=O)(=O)Cl | |
206.65 | |
206.65 | |
99% |
Chemical Identifiers
98-68-0 | |
206.65 | |
DTJVECUKADWGMO-UHFFFAOYSA-N | |
7401 | |
COC1=CC=C(C=C1)S(=O)(=O)Cl |
C7H7ClO3S | |
MFCD00007446 | |
4-methoxy-benzenesulfonyl chloride, 4-methoxybenzene-1-sulfonyl chloride, 4-methoxybenzenesulfonylchloride, 4-methoxybenzenesulphonyl chloride, 4-methoxyphenylsulfonyl chloride, benzenesulfonyl chloride, 4-methoxy, benzenesulfonyl chloride, p-methoxy, p-anisolesulfonyl chloride, p-anisylsulfonyl chloride, p-methoxyphenylsulfonyl chloride | |
4-methoxybenzenesulfonyl chloride |
Safety and Handling
GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Contact with water liberates toxic gas.
GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Store in a dry place. Store in a closed container.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger
EINECSNumber : 202-692-1