missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Methoxybenzenesulfonamide, 95%, ACROS Organics™

Manufacturer:  Acros Organics AC432910250

 View more versions of this product

Catalog No. AC432910250

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

4-Methoxybenzenesulfonamide
Glass bottle
109°C to 113°C
1129-26-6
100.0
C7H9NO3S
4-methoxybenzenesulphonamide, p-methoxybenzenesulfonamide, 4-methoxybenzene-1-sulfonamide, benzenesulfonamide, 4-methoxy, 4-methoxy-benzenesulfonamide, benzenesulfonamide, p-methoxy, benzenesulfonamide, p-methoxy-6ci,7ci,8ci, 3lka, p-anisolesulfonamide, acmc-1bvgf
COC1=CC=C(C=C1)S(=O)(=O)N
187.213
187.22
≥94.0%
Authentic
White
25g
94.0
95%
MFCD00025392
MSFQEZBRFPAFEX-UHFFFAOYSA-N
4-methoxybenzenesulfonamide
70789
Crystalline Powder

Chemical Identifiers

1129-26-6
187.213
MSFQEZBRFPAFEX-UHFFFAOYSA-N
70789
COC1=CC=C(C=C1)S(=O)(=O)N
C7H9NO3S
MFCD00025392
4-methoxybenzenesulphonamide, p-methoxybenzenesulfonamide, 4-methoxybenzene-1-sulfonamide, benzenesulfonamide, 4-methoxy, 4-methoxy-benzenesulfonamide, benzenesulfonamide, p-methoxy, benzenesulfonamide, p-methoxy-6ci,7ci,8ci, 3lka, p-anisolesulfonamide, acmc-1bvgf
4-methoxybenzenesulfonamide
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

SDS Documents