Learn More
4-Methoxybenzenediazonium tetrafluoroborate, 98%, Thermo Scientific Chemicals
$52.29 - $603.47
Chemical Identifiers
CAS | 459-64-3 |
---|---|
Molecular Formula | C7H7BF4N2O |
Molecular Weight (g/mol) | 221.95 |
MDL Number | MFCD00011897 |
InChI Key | CACFTKIREZJSIG-UHFFFAOYSA-J |
Synonym | 4-methoxybenzenediazonium tetrafluoroborate, 4-methoxybenzenediazoniumfluoroborate, 4-methoxybenzene diazonium fluoroborate, acmc-1aeuk, ksc236e8f, 4-methoxyphenyldiazonium tetrafluoroborate, p-methoxybenzenediazonium tetrafluoroborate, 4-methoxybenzene-1-diazonium; tetrafluoroboranuide |
PubChem CID | 517233 |
SMILES | [B+3].[F-].[F-].[F-].[F-].COC1=CC=C(C=C1)[N+]#N |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAH5582703
|
Thermo Scientific Chemicals
H5582703 |
1 g |
Each for $52.29
|
|
|||||
AAH5582706
|
Thermo Scientific Chemicals
H5582706 |
5 g |
Each for $164.00
|
|
|||||
AAH5582714
|
Thermo Scientific Chemicals
H5582714 |
25 g |
Each for $603.47
|
|
|||||
Description
4-Methoxybenzenediazonium tetrafluoroborate has been used in the preparation of azo coupled cyclic β-enaminones.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications4-Methoxybenzenediazonium tetrafluoroborate has been used in the preparation of azo coupled cyclic β-enaminones.
Solubility
Decompose in water
Notes
Store at -20°C. Light sensitive.
Chemical Identifiers
459-64-3 | |
221.95 | |
CACFTKIREZJSIG-UHFFFAOYSA-J | |
517233 |
C7H7BF4N2O | |
MFCD00011897 | |
4-methoxybenzenediazonium tetrafluoroborate, 4-methoxybenzenediazoniumfluoroborate, 4-methoxybenzene diazonium fluoroborate, acmc-1aeuk, ksc236e8f, 4-methoxyphenyldiazonium tetrafluoroborate, p-methoxybenzenediazonium tetrafluoroborate, 4-methoxybenzene-1-diazonium; tetrafluoroboranuide | |
[B+3].[F-].[F-].[F-].[F-].COC1=CC=C(C=C1)[N+]#N |
Specifications
459-64-3 | |
C7H7BF4N2O | |
1 g | |
4-methoxybenzenediazonium tetrafluoroborate, 4-methoxybenzenediazoniumfluoroborate, 4-methoxybenzene diazonium fluoroborate, acmc-1aeuk, ksc236e8f, 4-methoxyphenyldiazonium tetrafluoroborate, p-methoxybenzenediazonium tetrafluoroborate, 4-methoxybenzene-1-diazonium; tetrafluoroboranuide | |
CACFTKIREZJSIG-UHFFFAOYSA-J | |
4-methoxybenzenediazonium;tetrafluoroborate | |
517233 | |
98% |
142°C to 144°C | |
MFCD00011897 | |
UN3261 | |
Decompose in water | |
[B+3].[F-].[F-].[F-].[F-].COC1=CC=C(C=C1)[N+]#N | |
221.95 | |
221.95 | |
4-Methoxybenzenediazonium tetrafluoroborate |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
H302+H312+H332-H314
DOTInformation : Hazard Class: 8; Packaging Group: II
EINECSNumber : 207-296-2
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only