missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Methoxybenzenediazonium tetrafluoroborate, 98%

Manufacturer:  Alfa Aesar™ H5582714

 View more versions of this product

Catalog No. AAH5582714



Description

Description

Applications
4-Methoxybenzenediazonium tetrafluoroborate has been used in the preparation of azo coupled cyclic β-enaminones.

Solubility
Decompose in water

Notes
Store at -20°C. Light sensitive.
Specifications

Specifications

4-Methoxybenzenediazonium tetrafluoroborate
25g
C7H7BF4N2O
UN3261
Decompose in water
[B-](F)(F)(F)F.COC1=CC=C(C=C1)[N+]#N
221.95
221.95
142°C to 144°C
459-64-3
MFCD00011897
4-methoxybenzene diazonium fluoroborate, 4-methoxybenzene-1-diazonium; tetrafluoroboranuide, 4-methoxybenzenediazonium tetrafluoroborate, 4-methoxybenzenediazoniumfluoroborate, 4-methoxyphenyldiazonium tetrafluoroborate, acmc-1aeuk, ksc236e8f, p-methoxybenzenediazonium tetrafluoroborate
BYGWNWAFXUPZHI-UHFFFAOYSA-N
4-methoxybenzenediazonium;tetrafluoroborate
517233
98%

Chemical Identifiers

459-64-3
221.95
BYGWNWAFXUPZHI-UHFFFAOYSA-N
517233
[B-](F)(F)(F)F.COC1=CC=C(C=C1)[N+]#N
C7H7BF4N2O
MFCD00011897
4-methoxybenzene diazonium fluoroborate, 4-methoxybenzene-1-diazonium; tetrafluoroboranuide, 4-methoxybenzenediazonium tetrafluoroborate, 4-methoxybenzenediazoniumfluoroborate, 4-methoxyphenyldiazonium tetrafluoroborate, acmc-1aeuk, ksc236e8f, p-methoxybenzenediazonium tetrafluoroborate
4-methoxybenzenediazonium;tetrafluoroborate
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c

H302+H312+H332-H314

DOTInformation : Hazard Class: 8; Packaging Group: II

EINECSNumber : 207-296-2

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents