Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Methoxybenzaldehyde azine, 99%

$113.59 - $113.59

Chemical Identifiers

Chemical Name or Material 4-Methoxybenzaldehyde azine
CAS 2299-73-2
Molecular Formula C16H16N2O2
Formula Weight 268.32
MDL Number MFCD00025835
Beilstein 1079059
Synonym p-Anisaldehyde azine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1307606

 
alfa aesar™
L1307606
5g Each for $113.59
Description & Specifications

Specifications

Formula Weight 268.32
Assay 99%
CAS 2299-73-2
Synonym p-Anisaldehyde azine
Chemical Name or Material 4-Methoxybenzaldehyde azine
TSCA TSCA
Molecular Formula C16H16N2O2
MDL Number MFCD00025835
Beilstein 1079059
Melting Point 167° to 170°C
Quantity 5g