Learn More
4-Methoxybenzaldehyde, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A1536418
Description
4-Methoxybenzaldehyde is widely utilized in the fragrance and flavor industry. It finds application as an important intermediate in the synthesis of other organic compounds, perfumes and pharmaceuticals like antihistamines. It is also used in the preparation of agrochemicals, dyes and plastic additives. A solution of para-anisaldehyde with acid and ethanol is used as stain in thin layer chromatography (TLC), which provides easy identification of different compounds.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications4-Methoxybenzaldehyde is widely utilized in the fragrance and flavor industry. It finds application as an important intermediate in the synthesis of other organic compounds, perfumes and pharmaceuticals like antihistamines. It is also used in the preparation of agrochemicals, dyes and plastic additives. A solution of para-anisaldehyde with acid and ethanol is used as stain in thin layer chromatography (TLC), which provides easy identification of different compounds.
Solubility
Miscible with acetone, alcohol, ether, chloroform and benzene. Immiscible with water.
Notes
Air and Light Sensitive. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong bases, strong oxidizing agents and strong reducing agents.
Specifications
4-Methoxybenzaldehyde | |
0°C to 2°C | |
247°C to 249°C | |
98% | |
C8H8O2 | |
MFCD00003385 | |
471382 | |
14,663 | |
Miscible with acetone,alcohol,ether,chloroform and benzene. Immiscible with water. | |
COC1=CC=C(C=C1)C=O | |
136.15 | |
CHEBI:28235 | |
98% |
123-11-5 | |
1.121 | |
116°C (241°F) | |
Odorless | |
1.573 | |
50 g | |
Air and light sensitive | |
p-anisaldehyde, anisaldehyde, anisic aldehyde, p-methoxybenzaldehyde, 4-anisaldehyde, aubepine, p-anisic aldehyde, p-formylanisole, benzaldehyde, 4-methoxy, crategine | |
ZRSNZINYAWTAHE-UHFFFAOYSA-N | |
4-methoxybenzaldehyde | |
31244 | |
136.15 | |
Liquid |
Chemical Identifiers
123-11-5 | |
136.15 | |
ZRSNZINYAWTAHE-UHFFFAOYSA-N | |
31244 | |
4-methoxybenzaldehyde |
C8H8O2 | |
MFCD00003385 | |
p-anisaldehyde, anisaldehyde, anisic aldehyde, p-methoxybenzaldehyde, 4-anisaldehyde, aubepine, p-anisic aldehyde, p-formylanisole, benzaldehyde, 4-methoxy, crategine | |
CHEBI:28235 | |
COC1=CC=C(C=C1)C=O |
Safety and Handling
GHS H Statement
H302
Harmful if swallowed.
GHS P Statement
P264-P270-P301+P312-P330-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell. Rinse mouth.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Warning
EINECSNumber : 204-602-6
RTECSNumber : BZ2625000
TSCA : Yes
storageNote1 : Air sensitive
Recommended Storage : Ambient temperatures
RUO – Research Use Only