missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4′-Methoxyacetophenone 99.0+%, TCI America™

Manufacturer:  TCI America M0105500G

 View more versions of this product

Catalog No. M0105500G



Specifications

Specifications

4′-Methoxyacetophenone
White
500g
C9H10O2
4'-methoxyacetophenone, 4-acetylanisole, 4-methoxyacetophenone, 1-4-methoxyphenyl ethanone, p-methoxyacetophenone, acetanisole, novatone, linarodin, vananote, ethanone, 1-4-methoxyphenyl
CC(=O)C1=CC=C(C=C1)OC
150.177
CHEBI:86567
Crystal-Powder at 20°C
260°C
38°C
100-06-1
MFCD00008745
NTPLXRHDUXRPNE-UHFFFAOYSA-N
1-(4-methoxyphenyl)ethanone
7476
150.18
≥99.0% (GC)

Chemical Identifiers

100-06-1
150.177
NTPLXRHDUXRPNE-UHFFFAOYSA-N
7476
1-(4-methoxyphenyl)ethanone
C9H10O2
MFCD00008745
4'-methoxyacetophenone, 4-acetylanisole, 4-methoxyacetophenone, 1-4-methoxyphenyl ethanone, p-methoxyacetophenone, acetanisole, novatone, linarodin, vananote, ethanone, 1-4-methoxyphenyl
CHEBI:86567
CC(=O)C1=CC=C(C=C1)OC
Safety and Handling

Safety and Handling

EINECSNumber : (3)-2676

RTECSNumber : AM9240000

TSCA : Yes

Recommended Storage : Refrigerator

Documents