missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4'-Methoxyacetophenone, 99%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals A111620B

 View more versions of this product

Catalog No. AAA111620B



Description

Description

4'-Methoxyacetophenone is used as a component of perfumes and as chemical intermediate in the manufacture of pharmaceuticals, resins, flavouring agents. And also used to study ruthenium catalyzed step growth copolymerization of 4?-methoxyacetophenone with α,ω-dienes to yield copolymers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4′-Methoxyacetophenone is used as a component of perfumes and as chemical intermediate in the manufacture of pharmaceuticals, resins, flavouring agents. And also used to study ruthenium catalyzed step growth copolymerization of 4′-methoxyacetophenone with α,ω-dienes to yield copolymers.

Solubility
Insoluble in water.

Notes
Light Sensitive, Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications

Specifications

4'-Methoxyacetophenone
36°C to 40°C
258°C to 260°C
Characteristic
MFCD00008745
742313
4'-methoxyacetophenone, 4-acetylanisole, 4-methoxyacetophenone, 1-4-methoxyphenyl ethanone, p-methoxyacetophenone, acetanisole, novatone, linarodin, vananote, ethanone, 1-4-methoxyphenyl
NTPLXRHDUXRPNE-UHFFFAOYSA-N
1-(4-methoxyphenyl)ethanone
7476
150.18
100-06-1
1.08
138°C (280°F)
C9H10O2
1000 g
Light sensitive
Insoluble in water.
COC1=CC=C(C=C1)C(C)=O
150.18
CHEBI:86567
99%

Chemical Identifiers

100-06-1
150.18
NTPLXRHDUXRPNE-UHFFFAOYSA-N
7476
1-(4-methoxyphenyl)ethanone
C9H10O2
MFCD00008745
4'-methoxyacetophenone, 4-acetylanisole, 4-methoxyacetophenone, 1-4-methoxyphenyl ethanone, p-methoxyacetophenone, acetanisole, novatone, linarodin, vananote, ethanone, 1-4-methoxyphenyl
CHEBI:86567
COC1=CC=C(C=C1)C(C)=O
Safety and Handling

Safety and Handling

GHS H Statement
H301-H315
Toxic if swallowed.
Causes skin irritation.

P264b-P270-P301+P312-P330-P501c

H302

EINECSNumber : 202-815-9

RTECSNumber : AM9240000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

Promotions

Promotions

RUO – Research Use Only