missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
4'-Methoxyacetophenone, 98%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 152362500
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
4'-Methoxyacetophenone | |
100-06-1 | |
100.0 | |
138°C | |
97.5% min. (GC) | |
C9H10O2 | |
MFCD00008745 | |
08, 87 | |
Solubility in water: insoluble. Other solubilities: soluble in alcohol and ether | |
COC1=CC=C(C=C1)C(C)=O | |
150.18 | |
CHEBI:86567 | |
98% |
98% | |
97.5 | |
260.0°C | |
Authentic | |
Glass bottle | |
CH3OC6H4COCH3 | |
250g | |
4'-methoxyacetophenone, 4-acetylanisole, 4-methoxyacetophenone, 1-4-methoxyphenyl ethanone, p-methoxyacetophenone, acetanisole, novatone, linarodin, vananote, ethanone, 1-4-methoxyphenyl | |
NTPLXRHDUXRPNE-UHFFFAOYSA-N | |
1-(4-methoxyphenyl)ethanone | |
7476 | |
150.18 |
Safety and Handling
GHS H Statement
Harmful if swallowed.
GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Call a POISON CENTER or doctor/physician if you feel unwell.
Wash face,hands and any exposed skin thoroughly after handling.
GHS Signal Word: Warning
EINECSNumber : 202-815-9
RTECSNumber : AM9240000
TSCA : TSCA
RUO â Research Use Only