Please login to your online account to display your discounted pricing

4'-Methoxyacetanilide, 99+%, ACROS Organics™


Quantity 25g
Packaging Glass bottle

Manufacturer: fisher scientific acros  AC249710250

 View more versions of this product

Catalog No. AC249710250

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥99.0%
Physical Form Crystalline Powder
Formula Weight 165.19
Quantity 25g
Color Light beige
TSCA TSCA
RTECS Number AE8290000
EINECS Number 200-114-2
Melting Point 129° to 132°C
Merck Index 15, 50
CAS Min % 99.0
CAS Max % 100.0
Infrared Spectrum Authentic
Packaging Glass bottle
Health Hazard 3 GHS P Statement:
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Chemical Name or Material 4'-Methoxyacetanilide
Name Note 99+%
Assay Percent Range 99+%
Molecular Formula C9H11NO2
Linear Formula CH3C6H4NHC(O)CH3
MDL Number MFCD00014963
CAS 51-66-1
Synonym Methacetin
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.

Specifications

Chemical Name or Material 4'-Methoxyacetanilide
Name Note 99+%
CAS 51-66-1
CAS Min % 99.0
CAS Max % 100.0
Assay ≥99.0%
Assay Percent Range 99+%
Molecular Formula C9H11NO2
Linear Formula CH3C6H4NHC(O)CH3
Formula Weight 165.19
MDL Number MFCD00014963
Synonym Methacetin
Accessories

GHS P Statement:
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.

SDS & Safety Documents