missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4'-Methoxyacetanilide, 99+%, ACROS Organics™

$44.48 - $44.48

Chemical Identifiers

CAS 51-66-1
CAS Min % 99.0
CAS Max % 100.0
Assay Percent Range 99+%
Molecular Formula C9H11NO2
Linear Formula CH3C6H4NHC(O)CH3
Formula Weight 165.19
MDL Number MFCD00014963
Synonym Methacetin
Chemical Name or Material 4'-Methoxyacetanilide, 99+%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
AC249710250 SDS Fisher Scientific Acros
AC249710250
Glass bottle 25g Each for $44.48
Specifications Description & Specifications

Chemical Identifiers

51-66-1
100.0
C9H11NO2
165.19
Methacetin
≥99.0%
99.0
99+%
CH3C6H4NHC(O)CH3
MFCD00014963
4'-Methoxyacetanilide, 99+%

Specifications

165.19
25g
129° to 132°C
99.0
Glass bottle
Methacetin
CH3C6H4NHC(O)CH3
99+%
4'-Methoxyacetanilide, 99+%
Crystalline Powder
Light beige
15, 50
100.0
Authentic
51-66-1
C9H11NO2
MFCD00014963
≥99.0%

Safety and Handling

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement:
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

EINECSNumber : 200-114-2

RTECSNumber : AE8290000

TSCA : TSCA