Please login to your online account to display your discounted pricing

Alfa Aesar™ 4'-Methoxyacetanilide, 98+%

$92.66 - $386.36

Chemical Identifiers

Chemical Name or Material 4'-Methoxyacetanilide
CAS 51-66-1
Molecular Formula C9H11NO2
Formula Weight 165.19
MDL Number MFCD00014963
Beilstein 387887
Merck Index 14,51
Synonym p-Acetanisidine; N-(Methoxyphenyl)acetamide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1325330

 
alfa aesar™
A1325330
250g Each for $386.36

AAA1325318

 
alfa aesar™
A1325318
50g Each for $92.66
Description & Specifications

Specifications

Formula Weight 165.19
Chemical Name or Material 4'-Methoxyacetanilide
TSCA TSCA
Assay ≥98%
Molecular Formula C9H11NO2
MDL Number MFCD00014963
CAS 51-66-1
Beilstein 387887
Merck Index 14,51
Synonym p-Acetanisidine; N-(Methoxyphenyl)acetamide
Health Hazard 1 Danger
Melting Point 125° to 129°C
Quantity 250g
Health Hazard 2 GHS H Statement
H301
Toxic if swallowed.
Health Hazard 3 GHS P Statement
P264-P270-P301+P310-P321-P405-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Specific treatment (see label).
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
RTECS Number AE8290000

Applications
4′-Methoxyacetanilide is widely used in the preparation of disperse dyes. It is also used as an intermediate in organic synthesis.

Solubility
Slightly soluble in water.

Notes
Incompatible with strong oxidizing agents. Keep the container tightly closed in a dry and well-ventilated place.

Safety and Handling

Danger

GHS P Statement
P264-P270-P301+P310-P321-P405-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Specific treatment (see label).
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301
Toxic if swallowed.