missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Methoxy-trans-stilbene, TCI America™

$233.75 - $699.73

Chemical Identifiers

CAS 1694-19-5
Molecular Formula C15H14O
Molecular Weight (g/mol) 210.276
MDL Number MFCD00017178
InChI Key XWYXLYCDZKRCAD-BQYQJAHWSA-N
PubChem CID 1245920
ChEBI CHEBI:35094
IUPAC Name 1-methoxy-4-[(E)-2-phenylethenyl]benzene
SMILES COC1=CC=C(C=C1)C=CC2=CC=CC=C2
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M29645G View Documents TCI America
M29645G
5g Each for $699.73
 
M29641G View Documents TCI America
M29641G
1g Each for $233.75
 
Specifications

Chemical Identifiers

1694-19-5
210.276
XWYXLYCDZKRCAD-BQYQJAHWSA-N
CHEBI:35094
COC1=CC=C(C=C1)C=CC2=CC=CC=C2
C15H14O
MFCD00017178
1245920
1-methoxy-4-[(E)-2-phenylethenyl]benzene

Specifications

126°C
136°C
1694-19-5
MFCD00017178
COC1=CC=C(C=C1)C=CC2=CC=CC=C2
210.276
CHEBI:35094
Crystal-Powder at 20°C
4-Methoxy-trans-stilbene
White
5g
C15H14O
XWYXLYCDZKRCAD-BQYQJAHWSA-N
1-methoxy-4-[(E)-2-phenylethenyl]benzene
1245920
210.28
≥98.0% (GC)