Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Methoxy-TEMPO, free radical, 98+%

$69.46 - $265.19

Chemical Identifiers

Chemical Name or Material 4-Methoxy-TEMPO
CAS 95407-69-5
Name Note free radical
Molecular Formula C10H20NO2
Formula Weight 186.28
MDL Number MFCD00270334
Beilstein 4740399
Synonym 4-Methoxy-2,2,6,6-tetramethylpiperidinyloxy, free radical
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1591503

  SDS
alfa aesar™
L1591503
1g Each for $69.46

AAL1591506

  SDS
alfa aesar™
L1591506
5g Each for $265.19
Description & Specifications

Specifications

Formula Weight 186.28
Molecular Formula C10H20NO2
Assay ≥98%
Chemical Name or Material 4-Methoxy-TEMPO
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Synonym 4-Methoxy-2,2,6,6-tetramethylpiperidinyloxy, free radical
CAS 95407-69-5
Name Note free radical
MDL Number MFCD00270334
Health Hazard 1 Warning
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Melting Point 39° to 41°C
Beilstein 4740399
Quantity 1g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.