missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Methoxy-N-methylbenzylamine, 97%

Manufacturer:  Alfa Aesar™ H5577603

 View more versions of this product

Catalog No. AAH5577603



Specifications

Specifications

4-Methoxy-N-methylbenzylamine
702-24-9
MFCD04115407
1-4-methoxyphenyl-n-methyl-methanamine
CNCC1=CC=C(C=C1)OC
151.209
151.21
1g
C9H13NO
UN2735
AIJFPNKGGAPZFJ-UHFFFAOYSA-N
1-(4-methoxyphenyl)-N-methylmethanamine
485407
97%

Chemical Identifiers

702-24-9
151.209
AIJFPNKGGAPZFJ-UHFFFAOYSA-N
485407
CNCC1=CC=C(C=C1)OC
C9H13NO
MFCD04115407
1-4-methoxyphenyl-n-methyl-methanamine
1-(4-methoxyphenyl)-N-methylmethanamine
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

P201-P202-P261-P264b-P270-P271-P280i-P281-P301+P312-P302+P352-P304+P340-P305+P351+P338-P308+P313-P330-P332+P313-P362-P501c

H302-H315-H319-H335-H361

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.

TSCA : No

Recommended Storage : Keep cold; Store under Argon

Documents