missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Methoxy-N-(4-methylbenzyl)benzenesulfonamide, 97%

Manufacturer:  Alfa Aesar™ H5624503

 View more versions of this product

Catalog No. AAH5624503

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

4-Methoxy-N-(4-methylbenzyl)benzenesulfonamide
346696-69-3
MFCD01213791
PAIJRRXAEGQQNV-UHFFFAOYSA-N
4-methoxy-N-[(4-methylphenyl)methyl]benzenesulfonamide
743491
97%
1g
C15H17NO3S
4-methoxy-n-4-methylbenzyl benzenesulfonamide, 4-methoxy-n-4-methyl-benzyl-benzenesulfonamide, 4-methoxy-n-4-methylphenyl methyl benzenesulfonamide, n-4-methylbenzyl-4-methoxybenzenesulfonamide, 4-methoxyphenyl sulfonyl 4-methylphenyl methyl amine
CC1=CC=C(C=C1)CNS(=O)(=O)C2=CC=C(C=C2)OC
291.365
291.36

Chemical Identifiers

346696-69-3
291.365
PAIJRRXAEGQQNV-UHFFFAOYSA-N
743491
CC1=CC=C(C=C1)CNS(=O)(=O)C2=CC=C(C=C2)OC
C15H17NO3S
MFCD01213791
4-methoxy-n-4-methylbenzyl benzenesulfonamide, 4-methoxy-n-4-methyl-benzyl-benzenesulfonamide, 4-methoxy-n-4-methylphenyl methyl benzenesulfonamide, n-4-methylbenzyl-4-methoxybenzenesulfonamide, 4-methoxyphenyl sulfonyl 4-methylphenyl methyl amine
4-methoxy-N-[(4-methylphenyl)methyl]benzenesulfonamide
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

Documents