Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Methoxy-D-phenylalanine, 95%

Chemical Identifiers

Chemical Name or Material 4-Methoxy-D-phenylalanine
CAS 39878-65-4
Formula Weight 195.22
MDL Number MFCD00190961
Beilstein 3201704
Synonym H-D-Phe(4-OMe)-OH; (R)-2-Amino-3-(4-methoxyphenyl)propionic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6587203

 
alfa aesar™
H6587203
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6587206

 
alfa aesar™
H6587206
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6587214

 
alfa aesar™
H6587214
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 195.22
Chemical Name or Material 4-Methoxy-D-phenylalanine
Physical Form Solid
Assay 95%
Synonym H-D-Phe(4-OMe)-OH; (R)-2-Amino-3-(4-methoxyphenyl)propionic acid
Beilstein 3201704
Quantity 1g
Storage Note 1 2° to 8°C
CAS 39878-65-4
MDL Number MFCD00190961
Safety and Handling

2° to 8°C