Please login to your online account to display your discounted pricing

4-Methoxy-7-methyl-5H-furo[3,2-g][1]benzopyran-5-one, 97%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 4-Methoxy-7-methyl-5H-furo[3, 2-g][1]benzopyran-5-one
Name Note 97%
CAS 82-57-5
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C13H10O4
Formula Weight 230.22
MDL Number MFCD00005008
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC235030050

  SDS
thermo fisher scientific
235030050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥96.0%
Formula Weight 230.22
Physical Form Fine Crystalline Powder
Quantity 5g
Color Beige
RTECS Number LV1420000
EINECS Number 201-430-3
Melting Point 139° to 142°C
Merck Index 15, 10209
CAS Min % 96.0
CAS Max % 100.0
Synonym Visnagin
Infrared Spectrum Authentic
Packaging Glass bottle
Name Note 97%
MDL Number MFCD00005008
Health Hazard 2 GHS H Statement:
Harmful if swallowed.
Health Hazard 3 GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
CAS 82-57-5
Molecular Formula C13H10O4
Chemical Name or Material 4-Methoxy-7-methyl-5H-furo[3, 2-g][1]benzopyran-5-one
Assay Percent Range 97%
Safety and Handling

GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if swallowed.