missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Methoxy-6-methyl-2H-pyran-2-one, 97%

Manufacturer:  Alfa Aesar™ H61395MD

 View more versions of this product

Catalog No. AAH61395MD



Description

Description

Applications
4-Methoxy-6-methyl-2H-pyran-2-one is used as a model compound to study the assignment of ring proton resonances in 4-methoxy-6-nonyl-2H-pyran-2-one.

Solubility
Poorly soluble in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
Specifications

Specifications

4-Methoxy-6-methyl-2H-pyran-2-one
250mg
C7H8O3
2h-pyran-2-one, 4-methoxy-6-methyl
MTZAUZNQAMNFME-UHFFFAOYSA-N
4-methoxy-6-methylpyran-2-one
12651
Solid
83°C to 86°C
672-89-9
MFCD00006640
Poorly soluble in water.
CC1=CC(=CC(=O)O1)OC
140.138
140.14
97%

Chemical Identifiers

672-89-9
140.138
MTZAUZNQAMNFME-UHFFFAOYSA-N
12651
CC1=CC(=CC(=O)O1)OC
C7H8O3
MFCD00006640
2h-pyran-2-one, 4-methoxy-6-methyl
4-methoxy-6-methylpyran-2-one
Safety and Handling

Safety and Handling

EINECSNumber : 211-600-9

RTECSNumber : UQ1225500

TSCA : No

Recommended Storage : Ambient temperatures

Documents