missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Methoxy-4'-nitrobenzophenone, 97%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals B2272014

 View more versions of this product

Catalog No. AAB2272014



Description

Description

4-Methoxy-4'-nitrobenzophenone is used as an organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Methoxy-4′-nitrobenzophenone is used as an organic chemical synthesis intermediate.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Specifications

Specifications

4-Methoxy-4'-nitrobenzophenone
125°C to 127°C
MFCD00178348
4-methoxy-4'-nitrobenzophenone, 4-methoxyphenyl 4-nitrophenyl methanone, 4-methoxyphenyl-4-nitrophenyl methanone, methanone, 4-methoxyphenyl 4-nitrophenyl, 4-methoxy-phenyl-4-nitro-phenyl-methanone, acmc-1buw2, 4-methoxy-4-nitrobenzophenone, 4-methoxy-4?-nitrobenzophenone, 4-methoxy-4/'-nitrobenzophenone
COC1=CC=C(C=C1)C(=O)C1=CC=C(C=C1)[N+]([O-])=O
257.25
257.25
1151-94-6
C14H11NO4
25 g
DXVSAELNVPXMSQ-UHFFFAOYSA-N
(4-methoxyphenyl)-(4-nitrophenyl)methanone
347772
97%

Chemical Identifiers

1151-94-6
257.25
DXVSAELNVPXMSQ-UHFFFAOYSA-N
347772
COC1=CC=C(C=C1)C(=O)C1=CC=C(C=C1)[N+]([O-])=O
C14H11NO4
MFCD00178348
4-methoxy-4'-nitrobenzophenone, 4-methoxyphenyl 4-nitrophenyl methanone, 4-methoxyphenyl-4-nitrophenyl methanone, methanone, 4-methoxyphenyl 4-nitrophenyl, 4-methoxy-phenyl-4-nitro-phenyl-methanone, acmc-1buw2, 4-methoxy-4-nitrobenzophenone, 4-methoxy-4?-nitrobenzophenone, 4-methoxy-4/'-nitrobenzophenone
(4-methoxyphenyl)-(4-nitrophenyl)methanone
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only