Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Methoxy-4-methyl-2-pentanol, 98%

$60.66 - $518.74

Chemical Identifiers

Chemical Name or Material 4-Methoxy-4-methyl-2-pentanol
CAS 141-73-1
Molecular Formula C7H16O2
Formula Weight 132.2
MDL Number MFCD00046654
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2507806

 
alfa aesar™
B2507806
5g Each for $60.66

AAB2507814

 
alfa aesar™
B2507814
25g Each for $166.24

AAB2507822

 
alfa aesar™
B2507822
100g Each for $518.74
Description & Specifications

Specifications

Formula Weight 132.2
Chemical Name or Material 4-Methoxy-4-methyl-2-pentanol
Assay 98%
Molecular Formula C7H16O2
MDL Number MFCD00046654
CAS 141-73-1
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
RTECS Number SA7000000
Quantity 5g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.