Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Methoxy-3-(trifluoromethyl)phenol, 97%

$62.68 - $62.68

Chemical Identifiers

Chemical Name or Material 4-Methoxy-3-(trifluoromethyl)phenol
Molecular Formula C8H7F3O2
Formula Weight 192.14
MDL Number MFCD09832303
Synonym 5-Hydroxy-3-methoxybenzotrifluoride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH3152403

  SDS
alfa aesar™
H3152403
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH31524MD

  SDS
alfa aesar™
H31524MD
250mg Each for $62.68
Description & Specifications

Specifications

Formula Weight 192.14
Health Hazard 1 Danger
MDL Number MFCD09832303
Health Hazard 3 GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Synonym 5-Hydroxy-3-methoxybenzotrifluoride
Chemical Name or Material 4-Methoxy-3-(trifluoromethyl)phenol
Quantity 1g
Melting Point 45° to 47°C
Assay 97%
Molecular Formula C8H7F3O2
Health Hazard 2 GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Safety and Handling

Danger

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.