missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Methoxy-3-(trifluoromethyl)benzoyl chloride, 97%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals B2307203

 View more versions of this product

Catalog No. AAB2307203



Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

4-Methoxy-3-(trifluoromethyl)benzoyl chloride
C9H6ClF3O2
MFCD01631576
UN3265
4-methoxy-3-trifluoromethyl benzoyl chloride, 3-trifluoromethyl-p-anisoyl chloride, 5-chlorocarbonyl-2-methoxybenzotrifluoride, 4methoxy-3-trifluoromethylbenzoyl chloride, 4-methoxy-5-trifluoromethylbenzoyl chloride, 3-trifluoromethyl-4-methoxy-benzoyl chloride, 4-methoxy-3-trifluoromethyl benzoylchloride, 4-methoxy-3-trifluromethyl benzoyl chloride, 4-methoxy-3-trifluoromethyl-benzoyl chloride
COC1=CC=C(C=C1C(F)(F)F)C(Cl)=O
238.59
238.59
98187-18-9
1.515
1 g
Moisture sensitive
PBKYPTQBAQQVPT-UHFFFAOYSA-N
4-methoxy-3-(trifluoromethyl)benzoyl chloride
2775298
97%

Chemical Identifiers

98187-18-9
238.59
PBKYPTQBAQQVPT-UHFFFAOYSA-N
2775298
COC1=CC=C(C=C1C(F)(F)F)C(Cl)=O
C9H6ClF3O2
MFCD01631576
4-methoxy-3-trifluoromethyl benzoyl chloride, 3-trifluoromethyl-p-anisoyl chloride, 5-chlorocarbonyl-2-methoxybenzotrifluoride, 4methoxy-3-trifluoromethylbenzoyl chloride, 4-methoxy-5-trifluoromethylbenzoyl chloride, 3-trifluoromethyl-4-methoxy-benzoyl chloride, 4-methoxy-3-trifluoromethyl benzoylchloride, 4-methoxy-3-trifluromethyl benzoyl chloride, 4-methoxy-3-trifluoromethyl-benzoyl chloride
4-methoxy-3-(trifluoromethyl)benzoyl chloride
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c

H314-H335

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.

EINECSNumber : 000-000-0

TSCA : No

Recommended Storage : Ambient temperatures; Store under Nitrogen

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only