missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Methoxy-3-(trifluoromethyl)benzoyl chloride, 97%

Manufacturer:  Alfa Aesar B2307206

 View more versions of this product

Catalog No. AAB2307206


 

Specifications

Specifications

4-Methoxy-3-(trifluoromethyl)benzoyl chloride
97%
MFCD01631576
3-Trifluoromethyl-p-anisoyl chloride
PBKYPTQBAQQVPT-UHFFFAOYSA-N
4-methoxy-3-(trifluoromethyl)benzoyl chloride
2775298
98187-18-9
C9H6ClF3O2
UN3265
5g
COC1=C(C=C(C=C1)C(=O)Cl)C(F)(F)F
238.59
238.59

Chemical Identifiers

98187-18-9
238.59
PBKYPTQBAQQVPT-UHFFFAOYSA-N
2775298
C9H6ClF3O2
MFCD01631576
3-Trifluoromethyl-p-anisoyl chloride
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.

SDS Documents