Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Methoxy-3-(trifluoromethyl)benzonitrile, 97+%

Chemical Identifiers

Chemical Name or Material 4-Methoxy-3-(trifluoromethyl)benzonitrile
CAS 261951-87-5
Molecular Formula C9H6F3NO
Formula Weight 201.15
MDL Number MFCD01631575
UN Number UN3439
Synonym 3-Trifluoromethyl-p-anisonitrile
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2392303

  SDS
alfa aesar™
B2392303
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAB2392306

  SDS
alfa aesar™
B2392306
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 201.15
MDL Number MFCD01631575
Synonym 3-Trifluoromethyl-p-anisonitrile
Chemical Name or Material 4-Methoxy-3-(trifluoromethyl)benzonitrile
Health Hazard 1 Danger
CAS 261951-87-5
Health Hazard 3 GHS P Statement
P280h-P305+P351+P338-P309-P310
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
Molecular Formula C9H6F3NO
Assay ≥97%
Melting Point 98° to 100°C
Health Hazard 2 GHS H Statement
H311-H302-H332-H315-H319
Toxic in contact with skin.
Harmful if swallowed.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
Quantity 1g
UN Number UN3439
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
Safety and Handling

Danger

GHS P Statement
P280h-P305+P351+P338-P309-P310
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

GHS H Statement
H311-H302-H332-H315-H319
Toxic in contact with skin.
Harmful if swallowed.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.