Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Methoxy-3-(trifluoromethyl)benzoic acid, 97%

$76.50 - $252.45

Chemical Identifiers

Chemical Name or Material 4-Methoxy-3-(trifluoromethyl)benzoic acid
CAS 213598-09-5
Molecular Formula C9H7F3O3
Formula Weight 220.15
MDL Number MFCD01091014
Beilstein 8619237
Synonym 3-Trifluoromethyl-p-anisic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2273603

 
alfa aesar™
B2273603
1g Each for $76.50

AAB2273606

 
alfa aesar™
B2273606
5g Each for $252.45
Description & Specifications

Specifications

Formula Weight 220.15
Chemical Name or Material 4-Methoxy-3-(trifluoromethyl)benzoic acid
Assay 97%
Molecular Formula C9H7F3O3
MDL Number MFCD01091014
CAS 213598-09-5
Beilstein 8619237
Melting Point 169° to 171°C
Synonym 3-Trifluoromethyl-p-anisic acid
Quantity 1g
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.