Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Methoxy-3-(trifluoromethyl)benzaldehyde, 98%

$86.66 - $224.73

Chemical Identifiers

Chemical Name or Material 4-Methoxy-3-(trifluoromethyl)benzaldehyde
CAS 50823-87-5
Molecular Formula C9H7F3O2
Formula Weight 204.15
MDL Number MFCD01091012
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2612803

  SDS
alfa aesar™
H2612803
1g Each for $224.73

AAH26128MD

  SDS
alfa aesar™
H26128MD
250mg Each for $86.66
Description & Specifications

Specifications

Formula Weight 204.15
Chemical Name or Material 4-Methoxy-3-(trifluoromethyl)benzaldehyde
Assay 98%
Molecular Formula C9H7F3O2
MDL Number MFCD01091012
CAS 50823-87-5
Melting Point 38° to 40°C
Quantity 1g
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Solubility
Slightly soluble in water.

Notes
Air Sensitive.Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, air.

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.