missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Methoxy-3-(trifluoromethyl)aniline, 98+%

Manufacturer:  Alfa Aesar B2335109

 View more versions of this product

Catalog No. AAB2335109


 

Specifications

Specifications

4-Methoxy-3-(trifluoromethyl)aniline
≥98%
MFCD00043460
112°C (18mmHg)
57° to 61°C
10g
COC1=C(C=C(C=C1)N)C(F)(F)F
191.153
191.15
393-15-7
C8H8F3NO
UN2811
2723972
5-Amino-2-methoxybenzotrifluoride, 3-Trifluoromethyl-p-anisidine
CQJCPOVTPNWVBW-UHFFFAOYSA-N
4-methoxy-3-(trifluoromethyl)aniline
600705

Chemical Identifiers

393-15-7
191.153
CQJCPOVTPNWVBW-UHFFFAOYSA-N
600705
C8H8F3NO
MFCD00043460
5-Amino-2-methoxybenzotrifluoride, 3-Trifluoromethyl-p-anisidine
Safety and Handling

Safety and Handling

GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.

SDS Documents